missing translation for 'onlineSavingsMsg'
Learn More

Ammonium magnesium phosphate hydrate, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11355118
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Ammonium magnesium phosphate hydrate general application in pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7785-21-9
Molecular Formula H4MgNO4P
Molecular Weight (g/mol) 137.31
MDL Number MFCD00050358
InChI Key MXZRMHIULZDAKC-UHFFFAOYSA-L
Synonym unii-76yp0j930v, magnesium ammonium phosphate, ammonium magnesium phosphate hydrate, ammonium magnesium orthophosphate, phosphoric acid, ammonium magnesium salt, azanium magnesium phosphate, phosphoric acid, ammonium magnesium salt 1:1:1, magnesium 2+ ammonium phosphate, magnesium 2+ ion ammonium phosphate, ammonium magnesium phosphate nh4 mg po4
PubChem CID 178727
SMILES [NH4+].[Mg++].[O-]P([O-])([O-])=O

Specifications

Physical Form Powder
Quantity 50 g
Solubility Information Soluble in water.
IUPAC Name magnesium(2+) ammonium phosphate
Formula Weight 137.32 (Anhydrous)
Percent Purity 99.999%
Density 1.711 g/mL
Assay (metals basis)
Chemical Name or Material Ammonium magnesium phosphate hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'