missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium molybdate, 99.99%, (trace metal basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Quantity | 5 g |
Assay Percent Range | 99.99% |
Linear Formula | (NH4)2MoO4 |
Solubility Information | Solubility in water: soluble. Other solubilities: practically insoluble in alcohol |
Formula Weight | 196.01 |
Percent Purity | 99.99% |
Grade | Trace Metal Basis |
Packaging | Glass bottle |
Chemical Name or Material | Ammonium molybdate |
Safety and Handling

Product Identifier
- Ammonium molybdate
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ammonium molybdate, 99.99%, (trace metal basis), Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction