missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium molybdate tetrahydrate, ACS Reagent, 81.0-83.0% MoO3O basis, Honeywell Fluka™
Click to view available options
25 kg
Description
Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CAS | 12054-85-2 |
Quantity | 25 kg |
Molecular Formula | H6MoNO2 |
InChI Key | XCOMAVCHADVANQ-UHFFFAOYSA-O |
SMILES | [NH4+].O.[O--].[Mo] |
IUPAC Name | ammonium hydrate molybdenum oxidandiide |
Molecular Weight (g/mol) | 148.00 |
Chemical Name or Material | Ammonium molybdate tetrahydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'