missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Molybdate Tetrahydrate, Certified AR for Analysis, Fisher Chemical™

Product Code. 10725901
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg

H24MO7N6O24.4H2O, CAS Number-12054-85-2, 500g, White, 1235.86g/mol, 190 deg.C, 167059, Plastic powder jar, 5.0 to 5.5, Solid

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12054-85-2
Molecular Formula H6MoNO2
Molecular Weight (g/mol) 148.00
MDL Number 167059
InChI Key XCOMAVCHADVANQ-UHFFFAOYSA-O
IUPAC Name ammonium hydrate molybdenum oxidandiide
SMILES [NH4+].O.[O--].[Mo]

Specifications

Melting Point 190°C
Color White
pH 5.0 to 5.5
Physical Form Solid
Quantity 500 g
Formula Weight 1235.86g/mol
Packaging Plastic powder jar
compliance-icons
Product Identifier
  • Ammonium molybdate(VI) tetrahydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Ammonium heptamolybdate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.