missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Molybdate Tetrahydrate, puriss. p.a. ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
1 kg
25 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 100 g |
Linear Formula | (NH4)6Mo7O24 · 4H2O |
UN Number | NONH for all modes of transport |
Formula Weight | 1235.86g/mol |
Percent Purity | ≥99% (T) |
Grade | ACS/puriss. p.a. |
Packaging | Plastic Bottle |
Density | 2.498g/mL at 25°C |
Chemical Name or Material | Ammonium Molybdate Tetrahydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'