missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Nitrate, Honeywell Fluka™

Product Code. 15653230
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6484-52-2
Molecular Formula H4N2O3
Molecular Weight (g/mol) 80.04
MDL Number MFCD00011425
InChI Key DVARTQFDIMZBAA-UHFFFAOYSA-O
Synonym ammonium nitrate, nitram, ammonium nitricum, ammonium saltpeter, nitrate of ammonia, nitric acid ammonium salt, nitrato amonico, herco prills, german saltpeter, merco prills
PubChem CID 22985
ChEBI CHEBI:63038
IUPAC Name azanium;nitrate
SMILES [NH4+].[O-][N+]([O-])=O

Specifications

Quantity 2.5 kg
Linear Formula NH4NO3
UN Number 1942
Formula Weight 80.04g/mol
Percent Purity ≥98%
Grade ACS/puriss. p.a.
Packaging Plastic Bottle
Residue after Ignition ≤0.01% (as SO4)
Chemical Name or Material Ammonium Nitrate, Reag. Ph. Eur. reag. ISO
compliance-icons compliance-icons
Product Identifier
  • Ammonium nitrate
Signal Word
  • Warning
Hazard Category
  • Oxidising solid Category 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'