missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Oxalate Monohydrate, Certified AR for analysis, Fisher Chemical™
£48.05 - £325.00
Chemical Identifiers
CAS | 6009-70-7 |
---|---|
Molecular Formula | C2H10N2O5 |
Molecular Weight (g/mol) | 142.11 |
MDL Number | 149694 |
InChI Key | MSMNVXKYCPHLLN-UHFFFAOYSA-N |
IUPAC Name | oxalic acid diamine hydrate |
SMILES | N.N.O.OC(=O)C(O)=O |
Description
Ammonium Oxalate Monohydrate, Certified AR for analysis
Chemical Identifiers
6009-70-7 | |
142.11 | |
MSMNVXKYCPHLLN-UHFFFAOYSA-N | |
N.N.O.OC(=O)C(O)=O |
C2H10N2O5 | |
149694 | |
oxalic acid diamine hydrate |
Specifications
70°C | |
White | |
Solid | |
149694 | |
N.N.O.OC(=O)C(O)=O | |
142.11 | |
Plastic powder jar |
6009-70-7 | |
6.4 | |
C2H10N2O5 | |
MSMNVXKYCPHLLN-UHFFFAOYSA-N | |
oxalic acid diamine hydrate | |
142.11g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Ammonium oxalate monohydrate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : Ammonium oxalate, monohydrate, Ammoniumoxalate