missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Oxalate Monohydrate, Honeywell Fluka™
Click to view available options
Quantity:
250 g
500 g
1 kg
2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 500 g |
Linear Formula | (NH4)2C2O4 · H2O |
UN Number | NONH for all modes of transport |
Beilstein | 3759830 |
Formula Weight | 142.11g/mol |
Percent Purity | 99.5 to 101% |
Grade | ACS/puriss. p.a. |
Residue after Ignition | ≤0.02% (as SO4) |
Chemical Name or Material | Ammonium Oxalate Monohydrate, Reag. Ph. Eur. reag. ISO |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'