missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium persulfate, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
100 g
500 g
2 kg
5 kg
25 kg
Packaging:
Fibre drum
Glass Bottle
Plastic Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Oxidation reagent
Specifications
Specifications
Melting Point | 120.0°C |
Color | White |
Physical Form | Crystalline Powder and/or Crystals |
Quantity | 5 kg |
Assay Percent Range | 98+% |
Linear Formula | (NH4)2S2O8 |
Fieser | 01,952; 02,348; 03,238; 05,15; 06,20; 12,33 |
Merck Index | 15, 539 |
Solubility Information | Solubility in water: 582g/L (20°C), may decompose. |
Formula Weight | 228.19 |
Show More |
Safety and Handling
Product Identifier
- Ammonium persulfate
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 3
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- H272-May intensify fire; oxidiser.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'