missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Persulfate, Certified AR for Analysis, Fisher Chemical™
£39.65 - £61.33
Chemical Identifiers
CAS | 7727-54-0 |
---|---|
Molecular Formula | H8N2O8S2 |
Molecular Weight (g/mol) | 228.19 |
MDL Number | 3390 |
InChI Key | ROOXNKNUYICQNP-UHFFFAOYSA-N |
IUPAC Name | diammonium [(sulfonatoperoxy)sulfonyl]oxidanide |
SMILES | [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O |
Description
Chemical Identifiers
7727-54-0 | |
228.19 | |
ROOXNKNUYICQNP-UHFFFAOYSA-N | |
[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O |
H8N2O8S2 | |
3390 | |
diammonium [(sulfonatoperoxy)sulfonyl]oxidanide |
Specifications
120°C | |
White | |
Powder/Solid | |
3390 | |
[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O | |
228.19 | |
Plastic powder jar |
7727-54-0 | |
3.2 | |
H8N2O8S2 | |
ROOXNKNUYICQNP-UHFFFAOYSA-N | |
diammonium [(sulfonatoperoxy)sulfonyl]oxidanide | |
228.19g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Ammonium persulfate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 3
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- H272-May intensify fire; oxidiser.
Precautionary Statement
- P221-Take any precaution to avoid mixing with combustibles A§.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/A§.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/A§/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.