missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Phosphate Dibasic, puriss. p.a., Honeywell Fluka™
Puriss. p.a.,≥99.0% (T)
£20.36 - £126.07
Chemical Identifiers
CAS | 7783-28-0 |
---|---|
Molecular Formula | H9N2O4P |
Molecular Weight (g/mol) | 132.06 |
MDL Number | MFCD00010891 |
InChI Key | MNNHAPBLZZVQHP-UHFFFAOYSA-N |
Synonym | diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate |
PubChem CID | 24540 |
ChEBI | CHEBI:63051 |
IUPAC Name | diazanium;hydrogen phosphate |
SMILES | N.N.OP(O)(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15620920
|
Honeywell-Fluka
09840-50G |
50 g | N/A | ||||||
15610920
|
Honeywell-Fluka
09840-250G |
250 g |
£40.26
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15600920
|
Honeywell-Fluka
09840-1KG |
1 kg |
£126.07
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7783-28-0 | |
132.06 | |
MNNHAPBLZZVQHP-UHFFFAOYSA-N | |
24540 | |
diazanium;hydrogen phosphate |
H9N2O4P | |
MFCD00010891 | |
diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate | |
CHEBI:63051 | |
N.N.OP(O)(O)=O |
Specifications
7783-28-0 | |
(NH4)2HPO4 | |
NONH for all modes of transport | |
MNNHAPBLZZVQHP-UHFFFAOYSA-N | |
diazanium;hydrogen phosphate | |
24540 | |
132.06g/mol | |
puriss. p.a. |
H9N2O4P | |
MFCD00010891 | |
diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate | |
N.N.OP(O)(O)=O | |
132.06 | |
CHEBI:63051 | |
≥99% (T) | |
Ammonium Phosphate Dibasic |
Safety and Handling
missing translation for 'einecsNumber' : 231-987-8
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title