missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Phosphate, Monobasic, HPLC Grade, J.T. Baker™
570.00 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
1 kg
Packaging:
Wide Mouth Amber Glass Bottle
Description
- For Use in High Performance Liquid Chromatography
Specifications
CAS | 7722-76-1 |
Physical Form | Crystals |
Quantity | 1 kg |
Molecular Formula | H6NO4P |
MDL Number | MFCD00003396 |
Synonym | ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate |
InChI Key | LFVGISIMTYGQHF-UHFFFAOYSA-N |
SMILES | N.OP(O)(O)=O |
IUPAC Name | azanium;dihydrogen phosphate |
Molecular Weight (g/mol) | 115.03 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'