missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Sodium Phosphate Dibasic Tetrahydrate, ≥99.0% (NT), Honeywell Fluka™
Puriss. p.a.,≥99.0% (NT)
£32.96 - £102.39
Chemical Identifiers
CAS | 7783-13-3 |
---|---|
Molecular Formula | H13NNaO8P |
Molecular Weight (g/mol) | 209.067 |
MDL Number | MFCD00066970 |
InChI Key | IQTQISLCLWJRPM-UHFFFAOYSA-M |
Synonym | unii-k78u8wlv4h, ammonium sodium phosphate tetrahydrate, k78u8wlv4h, phosphorsalz, acmc-20ajp4, ammonium sodium phosphate tetrahydrate mi, sodium monoammonium monohydrogen phosphate tetrahydrate, phosphoric acid, monoammonium monosodium salt, tetrahydrate, salt of phosphorus microcosmic salt, sodium ammonium phosphate tetrahydrate |
PubChem CID | 56924540 |
IUPAC Name | azanium;sodium;hydrogen phosphate;tetrahydrate |
SMILES | [NH4+].O.O.O.O.OP(=O)([O-])[O-].[Na+] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7783-13-3 | |
209.067 | |
IQTQISLCLWJRPM-UHFFFAOYSA-M | |
56924540 | |
[NH4+].O.O.O.O.OP(=O)([O-])[O-].[Na+] |
H13NNaO8P | |
MFCD00066970 | |
unii-k78u8wlv4h, ammonium sodium phosphate tetrahydrate, k78u8wlv4h, phosphorsalz, acmc-20ajp4, ammonium sodium phosphate tetrahydrate mi, sodium monoammonium monohydrogen phosphate tetrahydrate, phosphoric acid, monoammonium monosodium salt, tetrahydrate, salt of phosphorus microcosmic salt, sodium ammonium phosphate tetrahydrate | |
azanium;sodium;hydrogen phosphate;tetrahydrate |
Specifications
7783-13-3 | |
H13NNaO8P | |
MFCD00066970 | |
unii-k78u8wlv4h, ammonium sodium phosphate tetrahydrate, k78u8wlv4h, phosphorsalz, acmc-20ajp4, ammonium sodium phosphate tetrahydrate mi, sodium monoammonium monohydrogen phosphate tetrahydrate, phosphoric acid, monoammonium monosodium salt, tetrahydrate, salt of phosphorus microcosmic salt, sodium ammonium phosphate tetrahydrate | |
[NH4+].O.O.O.O.OP(=O)([O-])[O-].[Na+] | |
209.067 | |
209.07g/mol | |
1.54g/cm3 |
Solid | |
NaNH4HPO4 · 4H2O | |
NONH for all modes of transport | |
IQTQISLCLWJRPM-UHFFFAOYSA-M | |
azanium;sodium;hydrogen phosphate;tetrahydrate | |
56924540 | |
≥99.0% (NT) | |
Ammonium sodium phosphate dibasic tetrahydrate |