missing translation for 'onlineSavingsMsg'
Learn More

Ammonium sulfate, 99.5%, for analysis, Thermo Scientific Chemicals

Chemical Identifiers

CAS 7783-20-2
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
Synonym ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
PubChem CID 6097028
ChEBI CHEBI:62946
SMILES N.N.OS(O)(=O)=O
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10424945
View Documents Promotion Details
Thermo Scientific Acros
205872500
250 g Plastic bottle N/A
N/A
10184780
View Documents Promotion Details
Thermo Scientific Acros
205870010
1 kg Plastic bottle N/A
N/A
10731424
View Documents
Thermo Scientific Acros
205870050
5 kg Plastic bottle N/A
N/A
11904251
View Documents
Thermo Scientific Acros
205870250
25 kg Fibre drum N/A
N/A
Edge
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7783-20-2
132.13
BFNBIHQBYMNNAN-UHFFFAOYSA-N
6097028
N.N.OS(O)(=O)=O
H8N2O4S
MFCD00003391
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
CHEBI:62946

Specifications

280.0°C
99.5%
(NH4)2SO4
15,552
Solubility in water: 754g/L. Other solubilities: insoluble in alcohol and acetone
N.N.OS(O)(=O)=O
132.13
CHEBI:62946
99.5%
0.01% max. (as SO4)
Ammonium sulfate
7783-20-2
H8N2O4S
MFCD00003391
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
BFNBIHQBYMNNAN-UHFFFAOYSA-N
diazanium;sulfate
6097028
132.14
Analytical
1.76
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

missing translation for 'einecsNumber' : 231-984-1

Product Identifier
  • Ammonium sulfate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only