missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium sulfate, 99+%, ACS reagent, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
500 g
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 280.0°C |
Color | Silver to Gray |
pH | 5.0 to 6.0 (5% soln. at 25°C) |
Physical Form | Granules, Fillings or Chips |
Quantity | 1 kg |
Assay Percent Range | 99+% |
Linear Formula | (NH4)2SO4 |
Merck Index | 15, 552 |
IUPAC Name | sulfuric acid diamine |
Formula Weight | 132.14 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'