Learn More
Ammonium sulfate, 99%, extra pure, Thermo Scientific Chemicals
£28.40 - £97.10
Chemical Identifiers
CAS | 7783-20-2 |
---|---|
Molecular Formula | H8N2O4S |
Molecular Weight (g/mol) | 132.13 |
MDL Number | MFCD00003391 |
InChI Key | BFNBIHQBYMNNAN-UHFFFAOYSA-N |
Synonym | ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate |
PubChem CID | 6097028 |
ChEBI | CHEBI:62946 |
IUPAC Name | sulfuric acid diamine |
SMILES | N.N.OS(O)(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11904261
|
Thermo Scientific Acros
434380010 |
1 kg | Plastic bottle |
£28.40
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13408588
|
Thermo Scientific Acros
434380050 |
5 kg | Plastic bottle |
£97.10
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7783-20-2 | |
132.13 | |
BFNBIHQBYMNNAN-UHFFFAOYSA-N | |
6097028 | |
sulfuric acid diamine |
H8N2O4S | |
MFCD00003391 | |
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate | |
CHEBI:62946 | |
N.N.OS(O)(=O)=O |
Specifications
280.0°C | |
White | |
Crystals | |
H8N2O4S | |
MFCD00003391 | |
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate | |
BFNBIHQBYMNNAN-UHFFFAOYSA-N | |
sulfuric acid diamine | |
6097028 | |
132.14 | |
Extra Pure |
7783-20-2 | |
5.0 to 6.0 | |
99% | |
(NH4)2SO4 | |
15,552 | |
Solubility in water: 754g/L. Other solubilities: insoluble in alcohol and acetone | |
N.N.OS(O)(=O)=O | |
132.13 | |
CHEBI:62946 | |
99% | |
Ammonium sulfate, Extra Pure |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
missing translation for 'einecsNumber' : 231-984-1
RUO – Research Use Only