missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Sulfate, Certified AR for Analysis, Fisher Chemical™
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Description
H8N2O4S, CAS Number-7783-20-2, 500g, Colorless, 132.13g/mol, 280 deg.C, 3391, Plastic powder jar, 5 to 6, Solid
Specifications
Specifications
Melting Point | 280°C |
Color | Colorless |
pH | 5 to 6 |
Physical Form | Solid |
Quantity | 1 kg |
Formula Weight | 132.13g/mol |
Packaging | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'