missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Sulfate, Certified AR for Analysis, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Unit Size:
1kg
2.5kg
500g
5kg
Description
H8N2O4S, CAS Number-7783-20-2, 500g, Colorless, 132.13g/mol, 280 deg.C, 3391, Plastic powder jar, 5 to 6, Solid
Specifications
Specifications
| Melting Point | 280°C |
| Color | Colorless |
| pH | 5 to 6 |
| Physical Form | Solid |
| Quantity | 500 g |
| Formula Weight | 132.13g/mol |
| Packaging | Plastic powder jar |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction