missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Sulfate, Fisher BioReagents
46.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Poly Bottle
Description
This enzyme-grade Ammonium Sulfate is suitable for use in the purification of proteins.
Specifications
Melting Point | 280°C |
Color | Colorless |
pH | 5 to 6 |
Physical Form | Solid |
Quantity | 1 kg |
Linear Formula | (NH4)2SO4 |
Merck Index | 15, 552 |
Formula Weight | 132.14 |
Percent Purity | ≥99.5% |
Identification | Pass Test |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'