missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium sulfate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99%, Honeywell Fluka™
Click to view available options
Quantity:
1 kg
5 kg
4 x 5 kg
50 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | >280°C (decomposition)(literature) |
CAS Max % | 100.0000% |
Quantity | 50 kg |
Linear Formula | HClO4 |
UN Number | NONH for all modes of transport |
Formula Weight | 132.14 |
Percent Purity | >99% |
Grade | ACS Reagent/Reagent ISO/Reagent Ph. Eur. |
Packaging | Fiberboard Box |
Residue after Ignition | ≤0.005% (as SO4) |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'