missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfate, purum p.a., Honeywell™

Product Code. 15600940
missing translation for 'orderingAttributeHoverText'
Quantity:
250 g
1 kg
5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Purum p.a.,≥99.0% (T)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-20-2
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
Synonym ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
PubChem CID 6097028
ChEBI CHEBI:62946
IUPAC Name diazanium;sulfate
SMILES N.N.OS(O)(=O)=O

Specifications

Quantity 5 kg
Linear Formula (NH4)2SO4
UN Number NONH for all modes of transport
Solubility Information Soluble in water, 0.5/10g/mL
Formula Weight 132.14g/mol
Percent Purity ≥99% (T)
Grade purum p.a.
Density 1.77g/mL at 25°C
Chemical Name or Material Ammonium Sulfate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.