missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium sulfate, saturated soln., Thermo Scientific™
Click to view available options
Quantity:
50 mL
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
CAS | 7783-20-2 |
Physical Form | Liquid |
Quantity | 50 mL |
Molecular Formula | H8N2O4S |
Linear Formula | (NH4)2SO4 |
MDL Number | MFCD00003391 |
Synonym | ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate |
InChI Key | BFNBIHQBYMNNAN-UHFFFAOYSA-N |
SMILES | N.N.OS(O)(=O)=O |
IUPAC Name | diazanium;sulfate |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'