missing translation for 'onlineSavingsMsg'
Learn More

Ammonium sulfide, 20% solution in water, Thermo Scientific Chemicals

£65.22 - £259.29

Chemical Identifiers

CAS 12135-76-1
Molecular Formula H8N2S
Molecular Weight (g/mol) 68.14
MDL Number MFCD00010892
InChI Key UXXGWUQNRMPNKH-UHFFFAOYSA-N
IUPAC Name diamine sulfane
SMILES N.N.S
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10490694
View Documents
Thermo Scientific Acros
426081000
100 mL Glass bottle
£65.22
100mL
Please to purchase this item. Need a web account? Register with us today!
10515524
View Documents
Thermo Scientific Acros
426080010
1 L Glass bottle
£259.29
1 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12135-76-1
68.14
UXXGWUQNRMPNKH-UHFFFAOYSA-N
N.N.S
H8N2S
MFCD00010892
diamine sulfane

Specifications

12135-76-1
(NH4)2S
32°C
Solubility in water: miscible
N.N.S
68.14
20.0 to 24.0% (Iodimetry)
1
Ammonium sulfide, 20% solution in water
H8N2S
MFCD00010892
15, 553
UXXGWUQNRMPNKH-UHFFFAOYSA-N
diamine sulfane
68.14
Clear yellow solution
1.0000g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Very toxic to aquatic life.
Contact with acids liberates toxic gas.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 235-223-4

Product Identifier
  • Ammonium sulfide, 20% solution in water

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
  • EUH031-Contact with acids liberates toxic gas.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only