missing translation for 'onlineSavingsMsg'
Learn More

Ammonium tetrafluoroborate, 99%, pure, Thermo Scientific Chemicals

Product Code. 10393003
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13826-83-0
Molecular Formula BF4H4N
Molecular Weight (g/mol) 104.84
MDL Number MFCD06245785
InChI Key PDTKOBRZPAIMRD-UHFFFAOYSA-O
Synonym ammonium tetrafluoroborate, ammonium fluoroborate, ammonium fluoborate, ammonium borofluoride, unii-c945z80o8x, ammonium tetrafluroborate, azanium tetrafluoroborate, arnmonium tetrafluoroborate, nh4bf4, ksc492c2p
PubChem CID 9964072
SMILES [NH4+].F[B-](F)(F)F

Specifications

Melting Point 230.0°C
Color Colorless
Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range 99%
Linear Formula NH4BF4
Fieser 05,17; 06,132
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ammonium hydride
IUPAC Name azanium;tetrafluoroborate
Formula Weight 104.84
Percent Purity 99%
Grade Pure
Packaging Plastic bottle
Chemical Name or Material Ammonium tetrafluoroborate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'