missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Thiocyanate, 0.1N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T. Baker™

Product Code. 15518344
Click to view available options
Quantity:
1 L
Packaging:
Narrow Mouth Polyethylene Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 1762-95-4
Quantity 1 L
Molecular Formula CH4N2S
Synonym ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt
InChI Key SOIFLUNRINLCBN-UHFFFAOYSA-N
SMILES C(#N)[S-].[NH4+]
IUPAC Name azanium;thiocyanate
Molecular Weight (g/mol) 76.117
PubChem CID 15666
Formula Weight 76.12
Grade BAKER ANALYZED™ Reagent
Packaging Narrow Mouth Polyethylene Bottle
Specific Gravity 1L = 1.10kg
Chemical Name or Material Ammonium Thiocyanate, 0.1N Volumetric Solution
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'