missing translation for 'onlineSavingsMsg'
Learn More

Ammonium thioglycolate, 70 wt.% solution in water, Thermo Scientific Chemicals

Ammonium thioglycolate, 68.0 to 72.0%, C2H7NO2S, CAS Number-5421-46-5, 7732-18-5

Brand:  Thermo Scientific Acros 384680010

57.20 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10521334

  • £57.49 / 1 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ammonium thioglycolate
70 wt.% solution in water
115.0°C
C2H7NO2S
1.2200g/mL
Solubility in water: completely soluble.
C(C(=O)[O-])S.[NH4+]
109.143
109.15
ammonium: positive, mercaptane: positive
1.22
1 L
Colorless
5421-46-5, 7732-18-5
-20.0°C
MFCD00137451
ammonium thioglycolate, ammonium mercaptoacetate, acetic acid, mercapto-, monoammonium salt, thiofaco a-50, ammonium thioglycollate, thioglycolic acid ammonium salt, usaf mo-2, unii-6p9e788vzz, thioglycollic acid, ammonium salt, ammonium 2-mercaptoacetate
ZZTCCAPMZLDHFM-UHFFFAOYSA-N
azanium;2-sulfanylacetate
21534
68.0 to 72.0%
Glass bottle
5.8 to 6.4
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

RUO – Research Use Only