missing translation for 'onlineSavingsMsg'
Learn More

Ammonium titanyl oxalate monohydrate, 98%, pure, Thermo Scientific Chemicals

Product Code. 10140452
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10580-03-7
Molecular Formula C4H8N2O9Ti
Molecular Weight (g/mol) 275.98
MDL Number MFCD00149620
InChI Key VBUNWMBOVJSSMH-UHFFFAOYSA-L
Synonym Ammonium bis(oxalato)oxotitanate(IV)
IUPAC Name diammonium oxotitaniumbis(ylium) dioxalate
SMILES [NH4+].[NH4+].O=[Ti++].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O

Specifications

Color White to Beige
Physical Form Crystalline Powder and/or Crystals
Quantity 100 g
Assay Percent Range 98%
Linear Formula (NH4)2TiO(C2O4)2·H2O
Merck Index 10,584
Solubility Information Solubility in water: soluble
Formula Weight 294.02
Percent Purity 98%
Grade Pure
Infrared Spectrum Authentic
Packaging Plastic bottle
Chemical Name or Material Ammonium titanyl oxalate monohydrate
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium titanyl oxalate monohydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.