Learn More
Ammonium vanadium oxide, 99% (metals basis), Thermo Scientific Chemicals
£57.90 - £176.00
Chemical Identifiers
CAS | 7803-55-6 |
---|---|
Molecular Formula | H4NO3V |
Molecular Weight (g/mol) | 116.98 |
MDL Number | MFCD00011430 |
InChI Key | YBVKNHXQSRDWAA-UHFFFAOYSA-M |
Synonym | ammonium metavanadate, ammonium vanadate v, ammonium monovanadate, unii-fl85px638g, vanadate vo31-, ammonium, vanadate vo31-, ammonium 1:1, ammonium vanadium oxide, ammoniummetavanadate, vanadic acid, hvo3 , ammonium salt, ammonium meta-vanadate |
PubChem CID | 516859 |
SMILES | N.O[V](=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11326108
|
Thermo Scientific Alfa Aesar
011091.30 |
250 g |
£57.90
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11316108
|
Thermo Scientific Alfa Aesar
011091.A1 |
1 kg |
£176.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ammonium vanadium oxide is used as a catalyst in organic synthesis, and as an analytical reagent in laboratories. It is employed in the preparation of vanadate. It acts as a sensor for the determination of N-acetyltransferase activity in humans. Further, it finds use in dyes, varnishes, indelible inks, drier for paints, and in photography. With copper carbonate, it has been shown to enhance the catalytic activities. It is an environmental-friendly catalyst to prepare alfa-hydroxyphosphonate derivatives in high yield through the reaction of aryl/heteroaryl aldehydes with triethylphosphonate derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7803-55-6 | |
116.98 | |
YBVKNHXQSRDWAA-UHFFFAOYSA-M | |
516859 |
H4NO3V | |
MFCD00011430 | |
ammonium metavanadate, ammonium vanadate v, ammonium monovanadate, unii-fl85px638g, vanadate vo31-, ammonium, vanadate vo31-, ammonium 1:1, ammonium vanadium oxide, ammoniummetavanadate, vanadic acid, hvo3 , ammonium salt, ammonium meta-vanadate | |
N.O[V](=O)=O |
Specifications
200°C (decomposition) | |
White | |
99% (Metal basis) | |
MFCD00011430 | |
Slightly soluble in water. Soluble in dilute ammonia and diethanolamine. Insoluble in ether,and alcohol. | |
N.O[V](=O)=O | |
116.98 | |
116.98 | |
2.33 g/cm3 |
7803-55-6 | |
Powder | |
H4NO3V | |
ammonium metavanadate, ammonium vanadate v, ammonium monovanadate, unii-fl85px638g, vanadate vo31-, ammonium, vanadate vo31-, ammonium 1:1, ammonium vanadium oxide, ammoniummetavanadate, vanadic acid, hvo3 , ammonium salt, ammonium meta-vanadate | |
YBVKNHXQSRDWAA-UHFFFAOYSA-M | |
vanadiumoylol amine | |
516859 | |
Off-white powder | |
Ammonium vanadium oxide |
Safety and Handling
P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P310-P304+P340-P305+P351+P338-P308+P313-P330-P501c
H301-H319-H332-H361d-H372
missing translation for 'dotInformation' : DOT Class: 6.1, Packaging Group: II
missing translation for 'einecsNumber' : 232-261-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only