missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amnionless/AMN Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-98805UV
This item is not returnable.
View return policy
Description
Amnionless/AMN Polyclonal specifically detects Amnionless/AMN in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Amnionless/AMN | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
amnionless homolog (mouse), PRO1028, protein amnionless, UNQ513/PRO1028, visceral endoderm-specific type 1 transmembrane protein | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human Amnionless/AMN. | |
0.1 mL | |
metabolism | |
81693 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction