missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amorolfine hydrochloride, 98%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
5 g
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Especificaciones
Melting Point | 221.0°C to 223.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (HPLC) |
Quantity | 250 mg |
Solubility Information | Solubility in water: 9g/L. Other solubilities: 28 mg/mL dmso, 71 mg/mL ethanol |
Formula Weight | 353.97 |
Percent Purity | 98% |
Chemical Name or Material | Amorolfine hydrochloride |
Seguridad y manipulación
Nombre del producto
- Amorolfine hydrochloride
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H411-Toxic to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P362-Take off contaminated clothing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'