missing translation for 'onlineSavingsMsg'
Learn More

AMOT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 16870-1-AP-150UL

 View more versions of this product

Product Code. 16826634

  • £349.00 / 150µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This antibody recognizes p130 only.

This gene belongs to the motin family of angiostatin binding proteins characterized by conserved coiled-coil domains and C-terminal PDZ binding motifs. The encoded protein is expressed predomitly in endothelial cells of capillaries as well as larger vessels of the placenta where it may mediate the inhibitory effect of angiostatin on tube formation and the migration of endothelial cells toward growth factors during the formation of new blood vessels. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications

Specifications

AMOT
Polyclonal
Unconjugated
AMOT
AMOT, angiomotin, KIAA1071
Rabbit
Antigen Affinity Chromatography
RUO
154796, 27494, 300289
-20°C
Liquid
Western Blot
0.34 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q4VCS5, Q8VHG2
AMOT
Peptide
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers