missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMOT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | AMOT |
---|---|
Concentration | 0.34 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16836634
|
Proteintech
16870-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16826634
|
Proteintech
16870-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody recognizes p130 only.
This gene belongs to the motin family of angiostatin binding proteins characterized by conserved coiled-coil domains and C-terminal PDZ binding motifs. The encoded protein is expressed predomitly in endothelial cells of capillaries as well as larger vessels of the placenta where it may mediate the inhibitory effect of angiostatin on tube formation and the migration of endothelial cells toward growth factors during the formation of new blood vessels. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
AMOT | |
Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
Q4VCS5, Q8VHG2 | |
154796, 27494, 300289 | |
Peptide | |
Primary | |
-20°C | |
AMOT |
0.34 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
AMOT, angiomotin, KIAA1071 | |
AMOT | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |