missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMPD, 0.2M buffer soln., pH 9.5, Thermo Scientific™
£45.73 - £60.50
Specifications
CAS | 115-69-5 |
---|---|
Color | Colorless |
Concentration | 0.2 M |
pH | 9.5 |
Physical Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15473599
|
Thermo Scientific Alfa Aesar
J60916.AE |
100 mL |
£45.73
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15483599
|
Thermo Scientific Alfa Aesar
J60916.AK |
250 mL |
£60.50
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
115-69-5 | |
0.2 M | |
Liquid | |
105.137 |
Colorless | |
9.5 | |
Odorless |
Safety and Handling
Recommended Storage : Keep cold