missing translation for 'onlineSavingsMsg'
Learn More

Amphotericin B, MP Biomedicals

Product Code. 11430892 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 mg
250 mg
500 mg
1 g
5 g
Unit Size:
100mg
1g
250mg
500mg
5g
This item is not returnable. View return policy

Product Code. 11430892

Brand: MP Biomedicals 0219504390

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1397-89-3
Molecular Formula C47H73NO17
Molecular Weight (g/mol) 924.09
MDL Number MFCD00877763
InChI Key APKFDSVGJQXUKY-ZNVUZQDLSA-N
Synonym Amphomoronal, Fungizone, Amphotericine B
PubChem CID 134129663
SMILES C[C@H]1O[C@@H](O[C@@H]2C[C@@H]3O[C@@](O)(C[C@H](O)[C@H]3C(O)=O)C[C@@H](O)C[C@@H](O)[C@H](O)CC[C@@H](O)C[C@@H](O)CC(=O)O[C@@H](C)[C@H](C)[C@H](O)[C@@H](C)\C=C/C=C\C=C/C=C\C=C/C=C\C=C/2)[C@@H](O)[C@@H](N)[C@@H]1O

Specifications

Type Amphotericin B
Physical Form Powder
Color Yellow
Quantity 500 mg
IUPAC Name (1S,3R,18S,19R,20R,21S,25R,27R,30R,31R,33S,35R,37S,38R)-3-[(2R,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy-19,25,27,30,31,33,35,37-octahydroxy-18,20,21-trimethyl-23-oxo-22,39-dioxabicyclo[33.3.1]nonatriaconta-4,6,8,10,12,14,16-heptaene-38-car
Formula Weight 924.1
Chemical Name or Material Amphotericin B, from Streptomyces sp.
compliance-icons
Product Identifier
  • Amphotericin B
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Amphotericin B
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.