missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amphotericin B, MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 mg
250 mg
500 mg
1 g
5 g
Unit Size:
100mg
1g
250mg
500mg
5g
Specifications
Specifications
| Type | Amphotericin B |
| Physical Form | Powder |
| Color | Yellow |
| Quantity | 1 g |
| IUPAC Name | (1S,3R,18S,19R,20R,21S,25R,27R,30R,31R,33S,35R,37S,38R)-3-[(2R,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy-19,25,27,30,31,33,35,37-octahydroxy-18,20,21-trimethyl-23-oxo-22,39-dioxabicyclo[33.3.1]nonatriaconta-4,6,8,10,12,14,16-heptaene-38-car |
| Formula Weight | 924.1 |
| Chemical Name or Material | Amphotericin B, from Streptomyces sp. |
Safety and Handling
Product Identifier
- Amphotericin B
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Amphotericin B
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction