missing translation for 'onlineSavingsMsg'
Learn More

Amphotericin B (Yellow Solution/250μg/mL), Fisher BioReagents

Product Code. 10746254
Click to view available options
Quantity:
20 mL
50 mL
Packaging:
Poly Bottle
Amphotericin B is effective against yeast and fungi. An antibiotic solution used in cell biology.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1397-89-3
Molecular Formula C47H73NO17
Molecular Weight (g/mol) 924.09
MDL Number MFCD00877763
InChI Key APKFDSVGJQXUKY-ZNVUZQDLSA-N
PubChem CID 134129663
IUPAC Name (1S,3R,18S,19R,20R,21S,25R,27R,30R,31R,33S,35R,37S,38R)-3-[(2R,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy-19,25,27,30,31,33,35,37-octahydroxy-18,20,21-trimethyl-23-oxo-22,39-dioxabicyclo[33.3.1]nonatriaconta-4,6,8,10,12,14,16-heptaene-38-car
SMILES C[C@H]1O[C@@H](O[C@@H]2C[C@@H]3O[C@@](O)(C[C@H](O)[C@H]3C(O)=O)C[C@@H](O)C[C@@H](O)[C@H](O)CC[C@@H](O)C[C@@H](O)CC(=O)O[C@@H](C)[C@H](C)[C@H](O)[C@@H](C)\C=C/C=C\C=C/C=C\C=C/C=C\C=C/2)[C@@H](O)[C@@H](N)[C@@H]1O

Specifications

Type Amphotericin B
Physical Form Liquid
Merck Index 15, 582
Packaging Poly Bottle
Formula Weight 924.1
Sterility Filter Sterilized
Color Yellow
Quantity 50 mL
Boiling Point 100°C
Chemical Name or Material Amphotericin B, 250μg/mL
Product Identifier
  • Amphotericin B Solution
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'