missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ampicillin Sodium Salt, MP Biomedicals™
Click to view available options
Quantity:
5 g
Specifications
Specifications
Type | Ampicillin Sodium Salt |
Physical Form | Powder |
CAS | 69-52-3 |
Molecular Formula | C16H21N3NaO4S |
Synonym | Aminobenzylpenicillin Sodium Salt |
InChI Key | BSFVNXCYXDYHOD-ZQDFAFASSA-N |
SMILES | [HH].CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)C.[Na] |
IUPAC Name | (2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium |
Molecular Weight (g/mol) | 374.411 |
PubChem CID | 131673879 |
Show More |
Safety and Handling
Product Identifier
- Ampicillin Sodium Salt
- Danger
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H317-May cause an allergic skin reaction.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'