missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMPK alpha 1 Antibody, Agarose, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NB600-396
Additional Details : Weight : 0.00970kg
Description
AMPK alpha 1 Polyclonal specifically detects AMPK alpha 1 in Human, Mouse samples. It is validated for Immunoprecipitation.Specifications
AMPK alpha 1 | |
Polyclonal | |
Agarose | |
PBS with 0.09% Sodium Azide | |
PRKAA1 | |
The immunogen recognized by this antibody maps to a region between residues 350 and 400 of human 5'-AMP-activated Protein Kinase, catalytic Alpha-1 chain using the numbering given in entry BAA36547.1 | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse | |
IgG |
Immunoprecipitation | |
0.5 mg/mL | |
Immunoprecipitation 1:10-1:500 | |
AMP -activate kinase alpha 1 subunit, AMP-activated protein kinase, catalytic, alpha-1,5'-AMP-activated protein kinase catalytic subunit alpha-1, AMPK, AMPK alpha 1,5'-AMP-activated protein kinase, catalytic alpha-1 chain, AMPK subunit alpha-1, AMPK1, AMPKa1, EC 2.7.11, EC 2.7.11.1, MGC33776, MGC57364, protein kinase, AMP-activated, alpha 1 catalytic subunit | |
Rabbit | |
64 kDa | |
0.1 mg | |
Apoptosis, Autophagy, Cancer, Hypoxia, MAP Kinase Signaling, mTOR Pathway, Signal Transduction, Translation Control | |
5562 | |
Store at 4C. Do not freeze. |
For Research Use Only