missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ AmpliTaq™ DNA Polymerase, LD (Low DNA)

Product Code. 10762697 Shop All Applied Biosystems Products
Click to view available options
Includes:
Buffer I
Buffer II
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10762697

Brand: Applied Biosystems™ N8080157

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Contains 250 units of AmpliTaq DNA Polymerase and Buffer I.

Applied Biosystems AmpliTaq DNA Polymerase, LD (Low DNA), is the same enzyme as AmpliTaq DNA Polymerase, but is further purified through a proprietary separation process to ensure that residual bacterial DNA sequences are substantially reduced.

This highly purified enzyme preparation ensures that non-target, false-positive PCR products will be effectively minimized when amplifying bacterial sequences. This is especially useful for low-copy-number PCR amplifications. The enzyme is quality-control tested to verify that fewer than 10 copies of bacterial 16S ribosomal RNA gene sequences are present in a standard 2.5 unit aliquot.

AmpliTaq DNA Polymerase, LD, is supplied with either 10X PCR Buffer I or with 10X PCR Buffer II and MgCl2 Solution.

Features

  • Minimizes non-specific PCR products when amplifying bacterial targets
  • Highly purified enzyme making it particularly useful for low copy number amplifications

Notes

  • See user's manual or package insert for limited label license and trademark information.

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 U/μL
Content And Storage • AmpliTaq DNA Polymerase, LD (5 U/μL), 25 μL
• 10X PCR Buffer I, 1.5 mL

Store at -15°C to -30°C.
Format Stand-alone enzyme
GC-Rich PCR Performance High
Polymerase AmpliTaq DNA Polymerase
Reaction Speed Standard
Exonuclease Activity 5' - 3'
Product Type DNA Polymerase
Purity or Quality Grade LD (Low DNA)
Quantity 250 units
Shipping Condition Dry Ice
For Use With (Application) Standard PCR
Fidelity (vs. Taq) 1X
Hot Start No
No. of Reactions 200 Reactions
Overhang 3'-A
Reaction Format Separate Components
Size (Final Product) 5 kb or less
Starting Material DNA
Includes Buffer I
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.