missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ AmpliTaq Gold™ DNA Polymerase with Gold Buffer and MgCl2

AmpliTaq Gold DNA Polymerase is a chemically modified form of AmpliTaq DNA Polymerase requiring thermal activation.

Brand:  Applied Biosystems™ 4311816

655.50 GBP valid until 2025-12-31
Use promo code "24357" to get your promotional price.


 View more versions of this product

Product Code. 10685095

  • £706.00 / 1000 units

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

• AmpliTaq Gold DNA Polymerase (5 U/μL), 1 x 200 μL
• GeneAmp 10X Gold Buffer, 4 x 1.5 mL
• 25 mM MgCl2, 4 x 1.5 mL

Store at -15°C to -30°C.
Low
Standard
DNA Polymerase
Dry Ice
5 U/μL
Built-In Hot Start
3'-A
5 kb or less
Tube
AmpliTaq Gold DNA Polymerase
5' - 3'
1000 Units
Hot-start PCR
1X
800
Separate Components
DNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2790246 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number4311816
Lot Number2790236 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number4311816
Lot Number2820434 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number4311816
Lot Number2790206 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number4311816
Lot Number2790221 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number4311816
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Applied Biosystems™ AmpliTaq Gold™ DNA Polymerase with Gold Buffer and MgCl2 > 1000 Units

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.