missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Ampule Breakers

Thermo Scientific Ampule Breakers provide a safe, convenient way to open 1 mL to 10 mL glass ampules.

Brand:  Thermo Scientific™ 28915

22.86 GBP valid until 2025-03-21
Use promo code "24111" to get your promotional price.


 View more versions of this product

Product Code. 15940372

  • £34.29 / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

This item is not returnable. View return policy

Description

Description

These single-use polyethylene sleeve ampule breakers help protect your fingers from glass shards when opening 1 mL to 10 mL glass ampules. The Thermo Scientific Ampule Breakers encapsulate the ampule top upon opening, making disposal of the glass top easy and safe.

• Easy to use single-use format
• Compatible with 1 mL to 10 mL glass ampules
• Protects against glass shards
• Helps prevent cross contamination

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Single-use
100 Breakers
100 breakers
Ampule Breaker
1-4 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Ampule Breakers > Ampule Breaker, 1-4 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.