Learn More
Labconco™ Ampules
Product Code. p-7111361 Shop All Labconco Corporation Products
Click to view available options
Capacity (Metric):
1 mL
5 mL
10 mL
Description:
Pre-scored, flat-bottom
Straight-sided
Unit Size:
100 ampoules
Pack of 100
Product Code. 10024084
Brand: Labconco™ 7571010
Only null left
Add to basket
Add to basket
Labconco Ampules are fabricated of highest quality borosilicate glass for strength and durability.
  • Constructed of high quality borosilicate glass for strength and durability
  • Supplied in packages of 100
Specifications
Capacity (Metric) 5 mL
Material Borosilicate Glass
Product Type Ampule
Description Pre-scored, flat-bottom