missing translation for 'onlineSavingsMsg'
Learn More

AMT hydrochloride, Tocris Bioscience™

Product Code. 18035692
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent, selective iNOS inhibitor

Potent, selective and reversible inhibitor of iNOS (IC50 = 3.6 nM; approximately 30 and 40 times selective over nNOS and eNOS respectively).

Chemical Identifiers

CAS 21463-31-0
Molecular Formula C5H10N2S
Molecular Weight (g/mol) 130.21
MDL Number MFCD00717539
InChI Key BATVOUKHGLKDGQ-SCSAIBSYSA-N
Synonym amt hydrochloride, 2-amino-5,6-dihydro-6-methyl-4h-1,3-thiazine, +/--amt hydrochloride, 2-amino-5,6-dihydro-6-methyl-4h-1,3-thiazine hydrochloride, 6-methyl-5,6-dihydro-4h-1,3-thiazin-2-amine hydrochloride, y-amt hydrochloride, ?-amt hydrochloride, c5h10n2s.hcl, inverted question mark-amt hydrochloride
PubChem CID 2733501
IUPAC Name (6R)-6-methyl-5,6-dihydro-4H-1,3-thiazin-2-amine
SMILES C[C@@H]1CCN=C(N)S1

Specifications

Quantity 10 mg
Formula Weight 166.67
Percent Purity >98%
Chemical Name or Material AMT hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'