missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Potent, selective and reversible inhibitor of iNOS (IC50 = 3.6 nM; approximately 30 and 40 times selective over nNOS and eNOS respectively).

Specifications
Specifications
Quantity | 50 mg |
Synonym | amt hydrochloride, 2-amino-5,6-dihydro-6-methyl-4h-1,3-thiazine, +/--amt hydrochloride, 2-amino-5,6-dihydro-6-methyl-4h-1,3-thiazine hydrochloride, 6-methyl-5,6-dihydro-4h-1,3-thiazin-2-amine hydrochloride, y-amt hydrochloride, ?-amt hydrochloride, c5h10n2s.hcl, inverted question mark-amt hydrochloride |
Formula Weight | 166.67 |
Percent Purity | >98% |
Chemical Name or Material | AMT hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
AMT hydrochloride, Tocris Bioscience™ >
Spot an opportunity for improvement?Share a Content Correction