missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMT hydrochloride, Tocris Bioscience™
Potent, selective iNOS inhibitor
£98.49 - £326.47
Chemical Identifiers
CAS | 21463-31-0 |
---|---|
Molecular Formula | C5H10N2S |
Molecular Weight (g/mol) | 130.21 |
MDL Number | MFCD00717539 |
InChI Key | BATVOUKHGLKDGQ-SCSAIBSYSA-N |
Synonym | amt hydrochloride, 2-amino-5,6-dihydro-6-methyl-4h-1,3-thiazine, +/--amt hydrochloride, 2-amino-5,6-dihydro-6-methyl-4h-1,3-thiazine hydrochloride, 6-methyl-5,6-dihydro-4h-1,3-thiazin-2-amine hydrochloride, y-amt hydrochloride, ?-amt hydrochloride, c5h10n2s.hcl, inverted question mark-amt hydrochloride |
PubChem CID | 2733501 |
IUPAC Name | (6R)-6-methyl-5,6-dihydro-4H-1,3-thiazin-2-amine |
SMILES | C[C@@H]1CCN=C(N)S1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18035692
|
Tocris Bioscience
0871/10 |
10 mg |
£98.49
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18074382
|
Tocris Bioscience
0871/50 |
50 mg |
£326.47
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potent, selective and reversible inhibitor of iNOS (IC50 = 3.6 nM; approximately 30 and 40 times selective over nNOS and eNOS respectively).Chemical Identifiers
21463-31-0 | |
130.21 | |
BATVOUKHGLKDGQ-SCSAIBSYSA-N | |
2733501 | |
C[C@@H]1CCN=C(N)S1 |
C5H10N2S | |
MFCD00717539 | |
amt hydrochloride, 2-amino-5,6-dihydro-6-methyl-4h-1,3-thiazine, +/--amt hydrochloride, 2-amino-5,6-dihydro-6-methyl-4h-1,3-thiazine hydrochloride, 6-methyl-5,6-dihydro-4h-1,3-thiazin-2-amine hydrochloride, y-amt hydrochloride, ?-amt hydrochloride, c5h10n2s.hcl, inverted question mark-amt hydrochloride | |
(6R)-6-methyl-5,6-dihydro-4H-1,3-thiazin-2-amine |
Specifications
21463-31-0 | |
MFCD00717539 | |
BATVOUKHGLKDGQ-SCSAIBSYSA-N | |
(6R)-6-methyl-5,6-dihydro-4H-1,3-thiazin-2-amine | |
2733501 | |
>98% |
C5H10N2S | |
amt hydrochloride, 2-amino-5,6-dihydro-6-methyl-4h-1,3-thiazine, +/--amt hydrochloride, 2-amino-5,6-dihydro-6-methyl-4h-1,3-thiazine hydrochloride, 6-methyl-5,6-dihydro-4h-1,3-thiazin-2-amine hydrochloride, y-amt hydrochloride, ?-amt hydrochloride, c5h10n2s.hcl, inverted question mark-amt hydrochloride | |
C[C@@H]1CCN=C(N)S1 | |
130.21 | |
166.67 | |
AMT hydrochloride |
Safety and Handling
Recommended Storage : Desiccate at +4°C