missing translation for 'onlineSavingsMsg'
Learn More

Amthamine dihydrobromide, Tocris Bioscience™

Product Code. 18054272
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Highly selective standard H2 agonist

Highly selective H2 agonist, slightly more potent than histamine itself. Only a weak antagonist at H3 and has no activity at H1 receptors. Induces vasodilation of cerebral arteries and decreases myogenic tone in vitro.

Chemical Identifiers

CAS 142457-00-9
Molecular Formula C6H13Br2N3S
Molecular Weight (g/mol) 319.059
InChI Key XFXNNOPUDSFVJE-UHFFFAOYSA-N
Synonym amthamine dihydrobromide, 5-thiazoleethanamine,2-amino-4-methyl, amthamine dihydrobromide hplc , solid, 2-amino-4-methyl-5-thiazoleethanamine dihydrobromide, 2-amino-5-2-aminoethyl-4-methylthiazole dihydrobromide, 5-2-aminoethyl-4-methyl-1,3-thiazol-2-amine-hydrogen bromide 1/2
PubChem CID 16218912
IUPAC Name 5-(2-aminoethyl)-4-methyl-1,3-thiazol-2-amine;dihydrobromide
SMILES CC1=C(SC(=N1)N)CCN.Br.Br

Specifications

Quantity 50 mg
Formula Weight 319.06
Percent Purity >99%
Chemical Name or Material Amthamine dihydrobromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'