Learn More
Thermo Scientific Chemicals Amylin, Human
Brand: Thermo Scientific Chemicals J66984.M¬
Code : NEW
Additional Details : CAS Number : 122384-88-7 Weight : 0.00001kg
Description
Amylin functions as part of the endocrine pancreas and contributes to glycemic control. It functions as a synergistic partner to insulin. The overall effect is to slow the rate of appearance (Ra) of glucose in the blood after eating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Human | |
-30°C to -10°C | |
3903.27 | |
1 mg |
HPLC | |
Solid | |
Amylin, human | |
RUO - research use only |
RUO – Research Use Only