Learn More
ANA-12, MedChemExpress
MedChemExpress ANA-12 is a potent and selective TrkB antagonist with IC50s of 45.6 nM and 41.1 μM for the high and low affinity sites, respectively.
Brand:  Medchem Express HY-12497
Encompass_Preferred
 View more versions of this product
Product Code. 16451427
  • £515.46 / 50mg
Explore more special offers
Description
Description
ANA-12 is a potent and selective TrkB antagonist with IC50s of 45.6 nM and 41.1 μM for the high and low affinity sites, respectively.
Specifications
Specifications
ANA-12
White
C22H21N3O3S
DMSO : 9.09 mg/mL (22.31 mM; Need ultrasonic) ∣Ethanol : < 1 mg/mL (ultrasonic) (insoluble)
407.49
99.3%
Solid
219766-25-3
Research
50 mg
O=C(C1=CC2=CC=CC=C2S1)NC3=CC=CC=C3C(NC4C(NCCCC4)=O)=O
407.49
Research
Neuroscience-Neuromodulation