missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Interscience™ Anabac Deodorizing Capsules
Placed in an autoclave, Anabac capsules provide a discreet way to perfume the laboratory.
Brand: Interscience™ 320200
This item is not returnable.
View return policy
Description
- Capsule dissolves during an autoclaving destruction cycle
- Capsule melts completely without leaving residue
- Eliminates unpleasant odors that can emerge from the autoclave
Specifications
Autoclave Capsule | |
Capsules | |
Cap | |
Sterile | |
Autoclavable | |
Single-use | |
Refreshes and Perfumes Air Where Autoclave Is Located | |
100 Capsules/Pk. |
Yes | |
Plastic | |
Apple | |
Clear Amber | |
Green, White | |
Non-filtered | |
Autoclave |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : 2-tert-butylcyclohexyl acetate, 2,6-dimethyl-7-octen-2-ol, cishex-3-en-1-ol, 2-propenyl hexanonate,
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction