missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Analog MultiTube Vortexer
Mix up to 50 samples at once.
Brand: Fisherbrand™ 80862898
1166.36 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Description
Mix up to 50 samples at once.
Vortexing action is created by holding the top tray of the vessel securely in place while the bottom tray moves freely in a defined, 3.6mm eccentric orbit.
- Powerful permanent, continuous-duty motor delivers quiet, rattle-free mixing
- Top plate adjusts along two stainless-steel posts; removable for easy sample access
- Rugged corrosion-resistant, stainless-steel base
- Suction-cup feet dampen motion and prevent sliding
- Orbit: 3.6mm
- Speed range: 1200-2400rpm
- Automatic timer (0 to 60 sec. for simple repeat procedures)
- 5 year warranty
Specifications
3.6 mm | |
230 V | |
Multi-Tube Vortexer | |
40.6 cm | |
24.1 cm | |
38.1 cm | |
19 kg | |
50 |
1200 to 2500 rpm | |
0.4 A | |
16 in. | |
9.5 in. | |
15 in. | |
42 lb. | |
50-place Foam rack, one tray pad set, 92 in. detachable three-wire cord and plug |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction